Orders LinkContactUsLinkComponent
VWR®, セルフ・ウインギング・モッピングシステム
VWR®, セルフ・ウインギング・モッピングシステム
General purpose high density sponge (406×127 mm), blue, for large volumes
Catalog # 129-2500
Supplier:   VWR Collection
VWR®, セルフ・ウインギング・モッピングシステム
Catalog # 129-2500
Supplier:  VWR Collection
VWR®, セルフ・ウインギング・モッピングシステム
General purpose high density sponge (406×127 mm), blue, for large volumes
Price
Inquire For Price
Log in for availability
1 * 50 items
Product Details & Documents
This mopping system can be used without a wringer. It is ideal for applying disinfectants to floors, walls and ceilings. The high performance foam for cleanrooms enables excellent, lasting liquid transfer.
  • Autoclavable; resistant to gamma rays and ETO
  • Resistant to the most widely available disinfectants and detergents
  • Sponges made from laminated polyester, for liquid absorption and highly delicate disinfection
  • Replacement foam mop heads are available with stainless steel or galvanised steel clips
Specifications
  • 説明:
    General purpose high density sponge (406×127 mm), blue, for large volumes
  • Seq:
    15