Sodium silicate in aqueous solution, TECHNICAL
Plastic bottle, 2,5 L
Supplier: VWR Chemicals
Sodium silicate in aqueous solution, TECHNICAL
Catalog # 28079.320
Supplier: VWR Chemicals
CAS Number: 1344-09-8
Jump to:
Product Details & Documents
Synonyms: Silicic acid sodium salt, Sodium silicate
- CAS Number:1344-09-8
- Formula:Na₂O(SiO₂)x·xH₂O
- MDL Number:MFCD00003492
- Density:1,34 g/cm³
- EINECS:215-687-4
- UN:3266
- ADR:8,III
Documents
Certificates
Search by Lot Number2,5L Plastic bottle for liquids DIN 45 UN X (ZPACK13962500) 25L Plastic drum for liquids DIN 61 (ZPACK13952500) 5L Plastic bottle for liquids DIN 45 UN X (ZPACK13951035)
Packaging Information
P20_VWRC28079.320_SPEC_EN.pdf P20_VWRC28079.363_SPEC_EN.pdf P20_VWRC28079.460_SPEC_EN.pdf spec_VWRC28079.320 spec_VWRC28079.320
Product specification
More
VWR Chemicals Big Packs and Bulks
Product Literature
Specifications
Specification Test Results
- Silica:25.6 - 27.6 %
- Sodium oxide:7.5 - 8.5 %
Specifications
- パックタイプ:Plastic bottle
- Pk:2,5 L
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



