Order Entry
Japan
Orders LinkContactUsLinkComponent
VWR®, コニカルストッパー、コルク

VWR®, コニカルストッパー、コルク

27 mm, 20 mm, 24 mm
Catalog # 217-1021
Supplier:   VWR Collection
VWR®, コニカルストッパー、コルク
Catalog # 217-1021
Supplier:  VWR Collection
VWR®, コニカルストッパー、コルク
VWR®, コニカルストッパー、コルク

27 mm, 20 mm, 24 mm

Price
Inquire For Price
Log in for availability
1 * 10 items
Product Details & Documents

Natural cork, A1 quality.

  • Lightweight, tough
  • Easy to drill holes into
  • Very economical
Specifications
  • Ø bottom:
    20 mm
  • Ø top:
    24 mm
  • 高さ:
    27 mm
  • Seq:
    11
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.