VWR®, コニカルストッパー、コルク
22 mm, 10 mm, 13 mm
Catalog # 217-1016
Supplier: VWR Collection
Jump to:
Product Details & Documents
Natural cork, A1 quality.
- Lightweight, tough
- Easy to drill holes into
- Very economical
Documents
Specifications
- Ø bottom:10 mm
- Ø top:13 mm
- 高さ:22 mm
- Seq:06