VWR®, コニカルストッパー、コルク
20 mm, 8 mm, 11 mm
Catalog # 217-1014
Supplier: VWR Collection
Jump to:
Product Details & Documents
Natural cork, A1 quality.
- Lightweight, tough
- Easy to drill holes into
- Very economical
Documents
Specifications
- Ø bottom:8 mm
- Ø top:11 mm
- 高さ:20 mm
- Seq:04