VWR®, 遠心分離フィルター
0,45, Purple
Catalog # 516-0236
Supplier: VWR Collection
Jump to:
Product Details & Documents
PP insert and receiver tube. MWCO filters are ideal to concentrate, purify and desalt peptides, proteins, oligonucleotides, DNA and RNA.
- Low binding, typical recovery greater than 90%
- Sealed membrane
- Fit rotors that accept 1,5 ml tubes
These 0,2 and 0,45 µm centrifugal filters are used for particulate removal, sample clarification or pre-filtration clean-up of large agarose or acrylamide gel debris prior to further purification with a MWCO device.
Fits rotors that accept 1,5 ml tubes
Specifications
- Colour code:Purple
- 細孔サイズ:0,45
- Seq:15
- Filtration area:0,28 cm²
- Cat. No.:516-0236
- Temperature:0...40 °C
- Sample volume:Max. 500 µl
- Dead volume:<5 µl
- Max. RCF:14000 ×g



