VWR®, 遠心分離フィルター
0,20 µm, Turquoise
516-0233
PP insert and receiver tube. MWCO filters are ideal to concentrate, purify and desalt peptides, proteins, oligonucleotides, DNA and RNA.
- Low binding, typical recovery greater than 90%
- Sealed membrane
- Fit rotors that accept 1,5 ml tubes
These 0,2 and 0,45 µm centrifugal filters are used for particulate removal, sample clarification or pre-filtration clean-up of large agarose or acrylamide gel debris prior to further purification with a MWCO device.
Fits rotors that accept 1,5 ml tubes
- Colour code:Turquoise
- 細孔サイズ:0,20 µm
- Seq:12
- Filtration area:0,28 cm²
- Cat. No.:516-0233
- Temperature:0...40 °C
- Sample volume:Max. 500 µl
- Dead volume:<5 µl
- Max. RCF:14000 ×g
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



