VWR®, ピペットチップ、スタンダード
5 - 200 µl, -, Yellow, Bulk
Catalog # 613-1056
Supplier: VWR Collection
Jump to:
Product Details & Documents
These universal tips are compatible with most micropipettes.
- Non sterile, autoclavable
- Smooth inside surface finish for minimum retention of dispensed liquid
Colour: Transparent (0,5 - 10 µl), yellow (2 - 200 µl and 5 - 200 µl), blue (200 - 1000 µl)
注文情報: Supplied loose or in racks holding 96 tips.
Specifications
- Packed:Bulk
- Sterile:-
- 容量:5 - 200 µl
- Seq:5
- Tip colour:Yellow