VWR®, ピペットチップ、スタンダード
0,5 - 10 µl, -, Transparent, 10 racks, 96 each
These universal tips are compatible with most micropipettes.
- Non sterile, autoclavable
- Smooth inside surface finish for minimum retention of dispensed liquid
Colour: Transparent (0,5 - 10 µl), yellow (2 - 200 µl and 5 - 200 µl), blue (200 - 1000 µl)
: Supplied loose or in racks holding 96 tips.
- Packed:10 racks, 96 each
- Sterile:-
- 容量:0,5 - 10 µl
- Seq:1
- Tip colour:Transparent