Orders LinkContactUsLinkComponent
VWR®, 秤量ボート
10 ml, 100 mm
  611-9196
VWR®, 秤量ボート
  611-9196
VWR®, 秤量ボート
VWR®, 秤量ボート
10 ml, 100 mm
1 * 3 items
Borosilicate 3.3 glass.
  • Ideal for weighing small samples
  • Powders can be tapped or rinsed through the tubular stem of the weighing boats into a receiving vessel
  • 容量:
    10 ml
  • 長さ:
    100 mm
  • Seq:
    03