VWR®, 秤量ボート
10 ml, 100 mm
611-9196
Borosilicate 3.3 glass.
- Ideal for weighing small samples
- Powders can be tapped or rinsed through the tubular stem of the weighing boats into a receiving vessel
- 容量:10 ml
- 長さ:100 mm
- Seq:03
Borosilicate 3.3 glass.