Orders LinkContactUsLinkComponent
VWR®, 秤量ボート
6 ml, 85 mm
  611-9195
VWR®, 秤量ボート
  611-9195
VWR®, 秤量ボート
VWR®, 秤量ボート
6 ml, 85 mm
1 * 3 items
Borosilicate 3.3 glass.
  • Ideal for weighing small samples
  • Powders can be tapped or rinsed through the tubular stem of the weighing boats into a receiving vessel
  • 容量:
    6 ml
  • 長さ:
    85 mm
  • Seq:
    02