VWR®, 秤量ボート
3 ml, 70 mm
Catalog # 611-9194
Supplier: VWR Collection
Jump to:
Product Details & Documents
Borosilicate 3.3 glass.
- Ideal for weighing small samples
- Powders can be tapped or rinsed through the tubular stem of the weighing boats into a receiving vessel
Documents
Specifications
- 容量:3 ml
- 長さ:70 mm
- Seq:01