VWR® 調剤ビン
Dispensing bin, 1 compartment, Blue, 115×155×170 mm
Catalog # 631-0720
Supplier: VWR Collection
Jump to:
Product Details & Documents
Acrylic.
Convenient, space saving bins are ergonomically designed with large angled openings to facilitate the removal of items. Bins feature non skid rubber feet for added bench top stability, as well as pre-drilled holes to allow for wall mounting (hardware included).
- Lid tilts back into locking position to remain open for repeated use
- Lid features an oversized lip to provide easy access, even with gloved hands
- Green bin is supplied with a removable insert divider to allow customisation of the interior (one or two compartments)
Documents
631-0720
Images 360
Specifications
- Colour:Blue
- 説明:Dispensing bin, 1 compartment
- W×D×H:115×155×170 mm