Order Entry
Japan
Orders LinkContactUsLinkComponent
 

VWR®, マイクロチューブラック、フローティング

White, 16 (4×4), 1,5/2,0 ml, 102×102×60 mm, Square
  211-0223
VWR®, マイクロチューブラック、フローティング
  211-0223
VWR®, マイクロチューブラック、フローティング
VWR®, マイクロチューブラック、フローティング

White, 16 (4×4), 1,5/2,0 ml, 102×102×60 mm, Square

 
 
   
1 * 4 items
 

PP.

  • Removable feet enable microtubes to be removed quickly and easily
  • Also suitable for cryotubes with up to 2 ml capacity
  • Stable, good chemical resistance

For maintaining microtubes with capacities from 0,5 to 2 ml at constant temperature in water baths. Racks float even when full. The tubes are held firmly in place in the frame and do not float. Handles are provided for easier insertion and removal from the water bath and for carrying. Not autoclavable.

 
  • For tubes:
    1,5/2,0 ml
  • Form:
    Square
  • 穴の数:
    16 (4×4)
  • Colour:
    White
  • Seq:
    03
  • W×D×H:
    102×102×60 mm