VWR®, クリーンルーム用モップ
Tubular knit mop
Catalog # 129-0147
Supplier: VWR Collection
Jump to:
Product Details & Documents
100% polyester string mops. Double clear poly bagged for cleanroom use. Good chemical sorbancy. Ideal for application of cleaning solutions on floors in critical environments.
- The sterile tubular knit mop, 129-0148, has a sterility assurance level of 10⁻⁶
- The ribbed tubular knit mop offers the best sorbancy with chemical
Suitable for use in Class 10, ISO 4, M2.5 environments or below.
Specifications
- 説明:Tubular knit mop
- Seq:01