Order Entry
Japan
Orders LinkContactUsLinkComponent
VWR®, ミニ機器ログブック

VWR®, ミニ機器ログブック

Mini equipment log book, Red, A5, 96
Catalog # 818-0108
Supplier:   VWR Collection
VWR®, ミニ機器ログブック
Catalog # 818-0108
Supplier:  VWR Collection
VWR®, ミニ機器ログブック
VWR®, ミニ機器ログブック

Mini equipment log book, Red, A5, 96

Price
Inquire For Price
Log in for availability
1 * 1 items
Product Details & Documents

'FLEX' covers with coating, for durability in laboratory environments. Acid-free, archive-safe paper (90 gsm).

  • Pages specifically designed for logging and recording equipment data
  • Tough coated cover

Format: A5 (14,8×21 cm) or A6 (10,5×14,8 cm), 96 numbered pages.

All books made in accordance with GLP, GMP and GCP guidelines.

Certifications: All books made in accordance with GLP, GMP and GCP guidelines.

Specifications
  • Paper size:
    A5
  • Colour:
    Red
  • 説明:
    Mini equipment log book
  • No. of pages:
    96
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.