Orders LinkContactUsLinkComponent
VWR®, 実験記録簿
Log book, balance calibration, Green, 168
  818-0104
VWR®, 実験記録簿
  818-0104
VWR®, 実験記録簿
VWR®, 実験記録簿
Log book, balance calibration, Green, 168
1 * 1 items
Imitation leather cover with coating, for durability in laboratory environments. Acid-free, archive-safe paper (90 gsm). Used to record and log data relating to laboratory equipment, temperature, balance calibration, reagents, animal maintenance and sample receipts.
  • All pages for logging documentation have specific column headings for easy data entry
  • Pages sewn in sections, books have round binding so that they lay flat
  • Waterproof and tough
All books made in accordance with GLP, GMP and GCP guidelines.
 : GLP, GMP and GCP guidelines.
  • Colour:
    Green
  • 説明:
    Log book, balance calibration
  • Seq:
    04
  • No. of pages:
    168