VWR®, 実験記録簿
Log book, balance calibration, Green, 168
818-0104
Imitation leather cover with coating, for durability in laboratory environments. Acid-free, archive-safe paper (90 gsm). Used to record and log data relating to laboratory equipment, temperature, balance calibration, reagents, animal maintenance and sample receipts.
- All pages for logging documentation have specific column headings for easy data entry
- Pages sewn in sections, books have round binding so that they lay flat
- Waterproof and tough
All books made in accordance with GLP, GMP and GCP guidelines.
: GLP, GMP and GCP guidelines.
Balance Calibration Log Book, Notes.pdf Balance Calibration Log Book, Page Sample.pdf Balance Calibration Log Book, User Data.pdf Reagent Log Book, Notes.pdf Reagent Log Book, Page Sample.pdf
- Colour:Green
- 説明:Log book, balance calibration
- Seq:04
- No. of pages:168