VWR®, 実験記録簿
Log book, laboratory equipment, Red, 96
818-0102
Imitation leather cover with coating, for durability in laboratory environments. Acid-free, archive-safe paper (90 gsm). Used to record and log data relating to laboratory equipment, temperature, balance calibration, reagents, animal maintenance and sample receipts.
- All pages for logging documentation have specific column headings for easy data entry
- Pages sewn in sections, books have round binding so that they lay flat
- Waterproof and tough
All books made in accordance with GLP, GMP and GCP guidelines.
Certifications: GLP, GMP and GCP guidelines.
Documents
Balance Calibration Log Book, Notes.pdf Balance Calibration Log Book, Page Sample.pdf Balance Calibration Log Book, User Data.pdf Reagent Log Book, Notes.pdf Reagent Log Book, Page Sample.pdf
Technical Drawing
Page Samples VWR Log Book 2016.pdf VWR Laboratory Notebooks VWR® for Calibrations VWR® for Weighing VWR® Lab bric-a-brac
Product Literature
TDS_VWR_LABORATORY_LOG_BOOKS_SPECIAL_PURPOSE.pdf
Product specification
- Colour:Red
- 説明:Log book, laboratory equipment
- Seq:01
- No. of pages:96