Order Entry
Japan
Orders LinkContactUsLinkComponent
VWR®, ラボラトリージャッキ用アクセサリー

VWR®, ラボラトリージャッキ用アクセサリー

Support, 245-0005, 245-0006, 245-0007, 245-0008, -
Catalog # 245-0014
Supplier:   VWR Collection
VWR®, ラボラトリージャッキ用アクセサリー
Catalog # 245-0014
Supplier:  VWR Collection
VWR®, ラボラトリージャッキ用アクセサリー
VWR®, ラボラトリージャッキ用アクセサリー

Support, 245-0005, 245-0006, 245-0007, 245-0008, -

Price
Inquire For Price
Log in for availability
1 * 1 items
Product Details & Documents

Extension panels: Aluminium, grey/black anodised. Increase the size of the work surface.

Stand plates: Aluminium, anodised grey/black. For securing to support stands.

Stand rods: Aluminium, anodised yellow. Can only be used in conjunction with the two extension panels 245-0017, 650 mm long, 12 mm diameter.

Stand fixture: Aluminium, anodised. For fixing laboratory racks to stand rods, etc.

Aluminium, anodised yellow. Can only be used in conjunction with the two extension panels 245-0017, 650 mm long, 12 mm diameter.

Documents

Specifications
  • For:
    245-0005, 245-0006, 245-0007, 245-0008
  • 説明:
    Support
  • Base:
    -