VWR®, Magnetic stirring bars, rectangular
35 mm, 6 mm
Catalog # 442-0488
Supplier: VWR Collection
Jump to:
Product Details & Documents
PTFE covered, strong Alnico V magnetic core. Smooth surface. Very effective stirrers, especially at low speeds.
- Excellent chemical resistance
- Temperature resistant, suitable for use at high and low temperatures (−200 to +280 °C)
Documents
Specifications
- 高さ:6 mm
- 長さ:35 mm