VWR®, マグネット式スターラーバー、オーバル
40 mm, 20 mm
442-0407
PTFE covered, Alnico V magnetic core. Smooth surface. Designed for use in round bottomed flasks.
- Excellent chemical resistance
- Temperature resistant, suitable for high and low temperature applications (−200 to +280 °C)
- 直径:20 mm
- 長さ:40 mm