VWR®, マグネット式スターラーバー、オーバル
35 mm, 16 mm
442-0406
PTFE covered, Alnico V magnetic core. Smooth surface. Designed for use in round bottomed flasks.
- Excellent chemical resistance
- Temperature resistant, suitable for high and low temperature applications (−200 to +280 °C)
- 直径:16 mm
- 長さ:35 mm