VWR®, マグネット式スターラーバー、オーバル
30 mm, 16 mm
Catalog # 442-0405
Supplier: VWR Collection
Jump to:
Product Details & Documents
PTFE covered, Alnico V magnetic core. Smooth surface. Designed for use in round bottomed flasks.
- Excellent chemical resistance
- Temperature resistant, suitable for high and low temperature applications (−200 to +280 °C)
Documents
Specifications
- 直径:16 mm
- 長さ:30 mm