Order Entry
Japan
Orders LinkContactUsLinkComponent
VWR®, マグネット式スターラーバー、オーバル

VWR®, マグネット式スターラーバー、オーバル

25 mm, 12 mm
Catalog # 442-0404
Supplier:   VWR Collection
VWR®, マグネット式スターラーバー、オーバル
Catalog # 442-0404
Supplier:  VWR Collection
VWR®, マグネット式スターラーバー、オーバル
VWR®, マグネット式スターラーバー、オーバル

25 mm, 12 mm

Price
Inquire For Price
Log in for availability
1 * 10 items
Product Details & Documents

PTFE covered, Alnico V magnetic core. Smooth surface. Designed for use in round bottomed flasks.

  • Excellent chemical resistance
  • Temperature resistant, suitable for high and low temperature applications (−200 to +280 °C)
Specifications
  • 直径:
    12 mm
  • 長さ:
    25 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.