VWR®, マグネット式スターラーバー、プレーン
75 mm, 13 mm
PTFE covered, strong Alnico V magnetic core. Smooth surface with rounded edges that intensify the stirring effect, even at low speeds.
- Excellent chemical resistance
- Temperature resistant, suitable for high and low temperature applications (−200 to +280 °C)
Documents
- 直径:13 mm
- 長さ:75 mm
- Seq:16