VWR®, マグネット式スターラーバー、プレーン
70 mm, 10 mm
Supplier: VWR Collection
Jump to:
Product Details & Documents
PTFE covered, strong Alnico V magnetic core. Smooth surface with rounded edges that intensify the stirring effect, even at low speeds.
- Excellent chemical resistance
- Temperature resistant, suitable for high and low temperature applications (−200 to +280 °C)
Specifications
- 直径:10 mm
- 長さ:70 mm
- Seq:15