VWR®, マグネット式スターラーバー、プレーン
50 mm, 10 mm
PTFE covered, strong Alnico V magnetic core. Smooth surface with rounded edges that intensify the stirring effect, even at low speeds.
- Excellent chemical resistance
- Temperature resistant, suitable for high and low temperature applications (−200 to +280 °C)
- 直径:10 mm
- 長さ:50 mm
- Seq:11