VWR®, マグネット式スターラーバー、レトリーバー
10 mm, 350 mm
Supplier: VWR Collection
Jump to:
Product Details & Documents
PTFE covered, strong Alnico V magnetic core. For the removal of stirring magnets from vessels of all kinds.
- Excellent chemical resistance
- Temperature resistant, suitable for use at high and low temperatures (−200 to +280 °C)
Specifications
- 長さ:350 mm
- Seq:005
- Stirrer Ø:10 mm