VWR®, マグネット式羽根型スターラー、マイクロ
20 mm, 13 mm, Container with max. Ø 14 - 15 mm
442-0497
PTFE covered, Alnico V magnetic core. Smooth surface. For stirring in test tubes, centrifuge tubes and micro vials. The wings can also be trimmed to size if needed.
- Excellent chemical resistance
- Temperature resistant, suitable for use at high and low temperatures (−200 to +280 °C)
- 直径:13 mm
- For:Container with max. Ø 14 - 15 mm
- 高さ:20 mm