VWR®, マグネティックスターリングディスク
70 mm, 37 mm
442-0492
PTFE covered, strong Alnico V magnetic core. The special shape with raised surface in the centre ensures very good agitation even at low speeds. Ideal for use in glass beakers.
- Excellent chemical resistance
- Temperature resistant, suitable for use at high and low temperatures (−200 to +280 °C)
- 直径:37 mm
- 長さ:70 mm