VWR®, マーカーペン
Marker pens, Green
Catalog # 811-0059
Supplier: VWR Collection
Jump to:
Product Details & Documents
Perfect for slides, sample bags and glassware.
- Write on glass, plastic, porcelain, film, rubber, wood and paper
- Permanent ink dries in seconds
Assorted colours: 6 black, 2 red, 2 blue, 2 green.
Specifications
- Colour:Green
- 説明:Marker pens
- Seq:04