Orders LinkContactUsLinkComponent
VWR®, マーカーペン
Marker pens, Blue
  811-0058
VWR®, マーカーペン
  811-0058
VWR®, マーカーペン
VWR®, マーカーペン
Marker pens, Blue
1 * 6 items
Perfect for slides, sample bags and glassware.
  • Write on glass, plastic, porcelain, film, rubber, wood and paper
  • Permanent ink dries in seconds
Assorted colours: 6 black, 2 red, 2 blue, 2 green.
  • Colour:
    Blue
  • 説明:
    Marker pens
  • Seq:
    03