VWR®, スライドフォルダー
Slide folder, White/black, Cardboard, 30
Catalog # 631-0690
Supplier: VWR Collection
Jump to:
Product Details & Documents
Slide trays hold 20 or 30 standard microscopy slides (76×26 mm).
- Thumb-cut for easy slide removal
- Horizontal compartments are recessed, protecting the specimens when the cover is closed
Colour: White (black rims)
Specifications
- No. of slides:30
- Colour:White/black
- 説明:Slide folder
- Material:Cardboard