VWR®, 輸送用綿棒、培地付き
Swabs for throat, vaginal, wound and skin use, Plastic/viscose, Amies liquid, White
Supplier: VWR Collection
Jump to:
Product Details & Documents
Collection and transport swabs for bacterial specimens with applicators and transport media.
- Bacteria will survive for 24 to 48 hours on the applicator and in the tube
- Tamper-proof seal, which turns white when open
- Medium risk for transient or short-term use
Suitable for surgical-invasive, short-term applications.
Class IIa - surgical-invasive, short-term application, for taking samples from natural orifices and surgical wounds.
: Class IIa.
: Individually wrapped aluminium pouches under nitrogen ensuring longer shelf life.
: *The product(s) on this page are outside the scope of the EU Directive 2019/904
Specifications
- 説明:Swabs for throat, vaginal, wound and skin use
- Seq:002
- Shaft/head:Plastic/viscose
- Transport medium:Amies liquid
- Cap colour:White