VWR®, 輸送用綿棒、培地付き
Swabs for male urethral use, Aluminium/viscose, Amies gel with charcoal, Orange
710-0435
Collection and transport swabs for bacterial specimens with applicators and transport media.
- Bacteria will survive for 24 to 48 hours on the applicator and in the tube
- Tamper-proof seal, which turns white when open
- Medium risk for transient or short-term use
Suitable for surgical-invasive, short-term applications.
Class IIa - surgical-invasive, short-term application, for taking samples from natural orifices and surgical wounds.
: Class IIa.
: Individually wrapped aluminium pouches under nitrogen ensuring longer shelf life.
: *The product(s) on this page are outside the scope of the EU Directive 2019/904
- 説明:Swabs for male urethral use
- Seq:009
- Shaft/head:Aluminium/viscose
- Transport medium:Amies gel with charcoal
- Cap colour:Orange