Orders LinkContactUsLinkComponent
VWR®, 輸送用綿棒、培地付き
VWR®, 輸送用綿棒、培地付き
Swabs for throat, wound and urogenital use, Plastic/viscose, Amies gel with charcoal, Black
Catalog # 710-0434
Supplier:   VWR Collection
VWR®, 輸送用綿棒、培地付き
Catalog # 710-0434
Supplier:  VWR Collection
VWR®, 輸送用綿棒、培地付き
VWR®, 輸送用綿棒、培地付き
Swabs for throat, wound and urogenital use, Plastic/viscose, Amies gel with charcoal, Black
Price
Inquire For Price
Log in for availability
1 * 50 items
Product Details & Documents
Collection and transport swabs for bacterial specimens with applicators and transport media.
  • Bacteria will survive for 24 to 48 hours on the applicator and in the tube
  • Tamper-proof seal, which turns white when open
  • Medium risk for transient or short-term use
Suitable for surgical-invasive, short-term applications.
Class IIa - surgical-invasive, short-term application, for taking samples from natural orifices and surgical  wounds.
 : Class IIa.
 : Individually wrapped aluminium pouches under nitrogen ensuring longer shelf life.
 : *The product(s) on this page are outside the scope of the EU Directive 2019/904
Specifications
  • 説明:
    Swabs for throat, wound and urogenital use
  • Seq:
    004
  • Shaft/head:
    Plastic/viscose
  • Transport medium:
    Amies gel with charcoal
  • Cap colour:
    Black