Orders LinkContactUsLinkComponent
VWR®, 輸送用綿棒、培地付き
VWR®, 輸送用綿棒、培地付き
Swabs with mini tip for male urethral use, Aluminium/viscose, Amies gel, Orange
Catalog # 710-0433
Supplier:   VWR Collection
VWR®, 輸送用綿棒、培地付き
Catalog # 710-0433
Supplier:  VWR Collection
VWR®, 輸送用綿棒、培地付き
VWR®, 輸送用綿棒、培地付き
Swabs with mini tip for male urethral use, Aluminium/viscose, Amies gel, Orange
Price
Inquire For Price
Log in for availability
1 * 50 items
Product Details & Documents
Collection and transport swabs for bacterial specimens with applicators and transport media.
  • Bacteria will survive for 24 to 48 hours on the applicator and in the tube
  • Tamper-proof seal, which turns white when open
  • Medium risk for transient or short-term use
Suitable for surgical-invasive, short-term applications.
Class IIa - surgical-invasive, short-term application, for taking samples from natural orifices and surgical  wounds.
Certifications: Class IIa.
包装: Individually wrapped aluminium pouches under nitrogen ensuring longer shelf life.
注意: *The product(s) on this page are outside the scope of the EU Directive 2019/904
Specifications
  • 説明:
    Swabs with mini tip for male urethral use
  • Seq:
    007
  • Shaft/head:
    Aluminium/viscose
  • Transport medium:
    Amies gel
  • Cap colour:
    Orange