Orders LinkContactUsLinkComponent
VWR®, 拡散器、T字型
VWR®, 拡散器、T字型
Spreaders, T-shaped, Blue, 10/peel pouch
Catalog # 612-2653
Supplier:   VWR Collection
VWR®, 拡散器、T字型
Catalog # 612-2653
Supplier:  VWR Collection
VWR®, 拡散器、T字型
VWR®, 拡散器、T字型
Spreaders, T-shaped, Blue, 10/peel pouch
Price
Inquire For Price
Log in for availability
1 * 1.000 items
Product Details & Documents
PS, sterile. Designed for spreading and dispersing liquids across the surface of agar culture plates. Used for performing bacterial (for example, CFU) plate counts on water, milk and other liquid samples.
  • Smooth, rounded surfaces to prevent cutting or gouging of agar
  • T-shaped to ensure even pressure is applied while spreading
Specifications
  • Packed:
    10/peel pouch
  • Colour:
    Blue
  • 説明:
    Spreaders, T-shaped