VWR®, 拡散器、T字型
Spreaders, T-shaped, Blue, 1/peel pouch
PS, sterile. Designed for spreading and dispersing liquids across the surface of agar culture plates. Used for performing bacterial (for example, CFU) plate counts on water, milk and other liquid samples.
- Smooth, rounded surfaces to prevent cutting or gouging of agar
- T-shaped to ensure even pressure is applied while spreading
- Packed:1/peel pouch
- Colour:Blue
- 説明:Spreaders, T-shaped