Orders LinkContactUsLinkComponent
VWR®, ショウジョウバエ用ストックボトル
PP, square bottom, 170 ml
  734-1260
VWR®, ショウジョウバエ用ストックボトル
  734-1260
VWR®, ショウジョウバエ用ストックボトル
VWR®, ショウジョウバエ用ストックボトル
PP, square bottom, 170 ml
1 * 500 items
PP, round or square bottom or PE, square bottom bottles.
  • Thin wall design for excellent visibility
  • Pliable top for easy removal of paper lid
  • Moulded-in food level graduation marks
Bottle 734-1259 features a wider edge for a more secure paper cap fit.
Bottle 734-1250 has a moulded indent near the bottom, which helps to secure media.
PP bottles are autoclavable, PE bottles are not.
 : *The product(s) on this page are outside the scope of the EU Directive 2019/904
  • 容量:
    170 ml
  • 説明:
    PP, square bottom
  • Seq:
    03