VWR®, ショウジョウバエ用ストックボトル
PP, round bottom, 170 ml
Catalog # 734-1249
Supplier: VWR Collection
Jump to:
Product Details & Documents
PP, round or square bottom or PE, square bottom bottles.
- Thin wall design for excellent visibility
- Pliable top for easy removal of paper lid
- Moulded-in food level graduation marks
Bottle 734-1259 features a wider edge for a more secure paper cap fit.
Bottle 734-1250 has a moulded indent near the bottom, which helps to secure media.
PP bottles are autoclavable, PE bottles are not.
注意: *The product(s) on this page are outside the scope of the EU Directive 2019/904
Specifications
- 容量:170 ml
- 説明:PP, round bottom
- Seq:01