Orders LinkContactUsLinkComponent
VWR®, ショウジョウバエ用ストックボトル
VWR®, ショウジョウバエ用ストックボトル
PP, round bottom, 170 ml
Catalog # 734-1249
Supplier:   VWR Collection
VWR®, ショウジョウバエ用ストックボトル
Catalog # 734-1249
Supplier:  VWR Collection
VWR®, ショウジョウバエ用ストックボトル
VWR®, ショウジョウバエ用ストックボトル
PP, round bottom, 170 ml
Price
Inquire For Price
Log in for availability
1 * 500 items
Product Details & Documents
PP, round or square bottom or PE, square bottom bottles.
  • Thin wall design for excellent visibility
  • Pliable top for easy removal of paper lid
  • Moulded-in food level graduation marks
Bottle 734-1259 features a wider edge for a more secure paper cap fit.
Bottle 734-1250 has a moulded indent near the bottom, which helps to secure media.
PP bottles are autoclavable, PE bottles are not.
注意: *The product(s) on this page are outside the scope of the EU Directive 2019/904
Documents
Specifications
  • 容量:
    170 ml
  • 説明:
    PP, round bottom
  • Seq:
    01