Order Entry
Japan
Orders LinkContactUsLinkComponent
VWR®, ラベル、クライオ

VWR®, ラベル、クライオ

Labels for 1,5 - 2 ml tubes, on roll, Red, 33×13 mm, On roll
Catalog # 817-5013
Supplier:   VWR Collection
VWR®, ラベル、クライオ
Catalog # 817-5013
Supplier:  VWR Collection
VWR®, ラベル、クライオ
VWR®, ラベル、クライオ

Labels for 1,5 - 2 ml tubes, on roll, Red, 33×13 mm, On roll

Price
Inquire For Price
Log in for availability
1 * 1.000 items
Product Details & Documents

The labels and adhesive have been designed to withstand temperatures from –196 to +121 °C. VWR® labels are ideal for all cryostorage, and can also be used in most incubators.

  • No need to laminate
  • Use on tubes, racks and boxes

Documents

Specifications
  • Colour:
    Red
  • 説明:
    Labels for 1,5 - 2 ml tubes, on roll
  • 寸法:
    33×13 mm
  • Packed:
    On roll
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.