Order Entry
Japan
Orders LinkContactUsLinkComponent
VWR®, ラベル、クライオ

VWR®, ラベル、クライオ

Labels for 1,5 - 2 ml tubes, on roll, Blue, 33×13 mm, On roll
Catalog # 817-5011
Supplier:   VWR Collection
VWR®, ラベル、クライオ
Catalog # 817-5011
Supplier:  VWR Collection
VWR®, ラベル、クライオ
VWR®, ラベル、クライオ

Labels for 1,5 - 2 ml tubes, on roll, Blue, 33×13 mm, On roll

Price
Inquire For Price
Log in for availability
1 * 1.000 items
Product Details & Documents

The labels and adhesive have been designed to withstand temperatures from –196 to +121 °C. VWR® labels are ideal for all cryostorage, and can also be used in most incubators.

  • No need to laminate
  • Use on tubes, racks and boxes

Documents

Specifications
  • Colour:
    Blue
  • 説明:
    Labels for 1,5 - 2 ml tubes, on roll
  • 寸法:
    33×13 mm
  • Packed:
    On roll