VWR®, ラベル、クライオ
Labels for 0,2 ml PCR tubes, on roll, White, 20×6 mm, On roll
Catalog # 817-5007
Supplier: VWR Collection
Jump to:
Product Details & Documents
The labels and adhesive have been designed to withstand temperatures from –196 to +121 °C. VWR® labels are ideal for all cryostorage, and can also be used in most incubators.
- No need to laminate
- Use on tubes, racks and boxes
Documents
VWR® Lab bric-a-brac
Product Literature
Specifications
- Colour:White
- 説明:Labels for 0,2 ml PCR tubes, on roll
- 寸法:20×6 mm
- Packed:On roll