VWR®, ラベル、クライオ
Printer labels for racks and boxes, suitable for Printer labelss, Laser, White, 67×25 mm
817-5003
The labels and adhesive have been designed to withstand temperatures from –196 to +121 °C. VWR® labels are ideal for all cryostorage, and can also be used in most incubators.
- No need to laminate
- Use on tubes, racks and boxes
- Printing method:Laser
- Colour:White
- 説明:Printer labels for racks and boxes, suitable for Printer labelss
- 寸法:67×25 mm