Orders LinkContactUsLinkComponent
VWR®, 使い捨て秤量皿
VWR®, 使い捨て秤量皿
110×29 mm
Catalog # 611-1377
Supplier:   VWR Collection
VWR®, 使い捨て秤量皿
Catalog # 611-1377
Supplier:  VWR Collection
VWR®, 使い捨て秤量皿
VWR®, 使い捨て秤量皿
110×29 mm
Price
Inquire For Price
Log in for availability
1 * 50 items
Product Details & Documents
Aluminium, light gauge.
  • Flanged edge with curled lip for increased stability
  • Contaminant- and oil-free
  • Stackable
Ideal for weighing, sampling, dispensing, storage, food processing, evaporating or general laboratory use.
Specifications
  • Int.Ø×H:
    110×29 mm
  • Seq:
    04