Orders LinkContactUsLinkComponent
VWR®, 使い捨て秤量皿
180×25 mm
  611-1375
VWR®, 使い捨て秤量皿
  611-1375
VWR®, 使い捨て秤量皿
VWR®, 使い捨て秤量皿
180×25 mm
1 * 50 items
Aluminium, light gauge.
  • Flanged edge with curled lip for increased stability
  • Contaminant- and oil-free
  • Stackable
Ideal for weighing, sampling, dispensing, storage, food processing, evaporating or general laboratory use.
  • Int.Ø×H:
    180×25 mm
  • Seq:
    05