VWR®, 使い捨て秤量皿
180×25 mm
611-1375
Aluminium, light gauge.
- Flanged edge with curled lip for increased stability
- Contaminant- and oil-free
- Stackable
Ideal for weighing, sampling, dispensing, storage, food processing, evaporating or general laboratory use.
- Int.Ø×H:180×25 mm
- Seq:05