VWR®, 使い捨て秤量皿
44×13 mm
Catalog # 611-1374
Supplier: VWR Collection
Jump to:
Product Details & Documents
Aluminium, light gauge.
- Flanged edge with curled lip for increased stability
- Contaminant- and oil-free
- Stackable
Ideal for weighing, sampling, dispensing, storage, food processing, evaporating or general laboratory use.
Documents
VWR® for Weighing
Product Literature
Specifications
- Int.Ø×H:44×13 mm
- Seq:01