VWR®, ビーカーカップ、使い捨て、製
50 ml, 50 mm
611-1366
PS.
- Suitable for use in various sample dispensers
- Beakers with capacity of 5 and 20 ml are recommended for use in flame photometric analyses
Economical, multipurpose beaker for a very wide range of uses.
- 容量:50 ml
- 高さ:50 mm
- Seq:05



