Order Entry
Japan
Orders LinkContactUsLinkComponent
VWR®, ビーカーカップ、使い捨て、製

VWR®, ビーカーカップ、使い捨て、製

20 ml, 32 mm
Catalog # 611-1365
Supplier:   VWR Collection
VWR®, ビーカーカップ、使い捨て、製
Catalog # 611-1365
Supplier:  VWR Collection
VWR®, ビーカーカップ、使い捨て、製
VWR®, ビーカーカップ、使い捨て、製

20 ml, 32 mm

Price
Inquire For Price
Log in for availability
1 * 500 items
Product Details & Documents

PS.

  • Suitable for use in various sample dispensers
  • Beakers with capacity of 5 and 20 ml are recommended for use in flame photometric analyses

Economical, multipurpose beaker for a very wide range of uses.

Specifications
  • 容量:
    20 ml
  • 高さ:
    32 mm
  • Seq:
    04
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.